Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIT1 Antibody (SIT-01), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB500486549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500486549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500486549 Supplier Novus Biologicals Supplier No. NB500486JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIT1 Monoclonal specifically detects SIT1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SIT1
Applications Flow Cytometry
Classification Monoclonal
Clone SIT-01
Conjugate Janelia Fluor 549
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1
Gene Symbols SIT1
Host Species Mouse
Immunogen Bacterially produced recombinant intracellular fragment of human SIT.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27240
Test Specificity The antibody SIT-01 reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.