Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Sivelestat sodium salt
GREENER_CHOICE

Catalog No. 353510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-351-0 10 mg
35-355-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-351-0 Supplier Tocris Bioscience™ Supplier No. 3535/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective leukocyte elastase inhibitor

Selective leukocyte elastase inhibitor (IC50 = 44 nM) that displays no activity at a range of other proteases. Inhibits NF-κB activation and LTB4-induced neutrophil transmigration in vitro. Significantly attenuates ischemia-induced spinal cord injury, decreases serum cytokine levels and reduces acute inflammatory lung injury in vivo.

Chemical Identifiers

CAS 150374-95-1
Molecular Formula C20H21N2NaO7S

Specifications

Quantity 10 mg
Formula Weight 456.44
Purity >98%
Molecular Weight (g/mol) 456.445
Chemical Name or Material Sivelestat sodium salt
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.