Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX1 Antibody (CL0185), Novus Biologicals™
SDP

Catalog No. NB395383 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395383 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB395383 Supplier Novus Biologicals Supplier No. NBP25287325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIX1 Monoclonal antibody specifically detects SIX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown

Specifications

Antigen SIX1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Monoclonal
Clone CL0185
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated
Formulation PBS (pH 7.2), 40% Glycerol
Gene Alias BOS3, deafness, autosomal dominant 23, DFNA23, homeobox protein SIX1, sine oculis homeobox (Drosophila) homolog 1, Sine oculis homeobox homolog 1, sine oculis homeobox homolog 1 (Drosophila), SIX homeobox 1, TIP39
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: CFKEKSRGVLREWYAHNPYPSPREKRELAEATGLTTTQVSNWFKNRRQRDRAAEAKERENTENNNSSSNKQNQLSPLEGGKPLMSSSEEEFSPPQSPDQNSVLLLQGNMGHARSSNYSLPGLTASQPSHGLQTHQHQLQDS
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6495
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.