Learn More
SIX3 Polyclonal Antibody, Rockland™

Guinea Pig Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | SIX3 |
---|---|
Concentration | 0.65 mg/mL |
Applications | ELISA, Immunofluorescence, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0494
![]() ![]() |
Rockland Immunochemicals
200201A26 |
50 μg |
Each for $636.24
|
|
|||||
50-105-7532
![]() ![]() |
Rockland Immunochemicals
200201A26S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with mouse Six3. Cross-reactivity with Six3 from other sources has not been determined.
This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.Specifications
SIX3 | |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Guinea Pig | |
Mouse | |
Q62233 | |
20473 | |
This Protein A purified antibody was prepared from whole guinea pig serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of mouse Six3 protein. | |
-20°C, Avoid Freeze/Thaw Cycles | |
Six3 |
0.65 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
BOS_11581; E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; Sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta | |
Six3 | |
Primary | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.