Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX6 Antibody, Novus Biologicals™
SDP

Catalog No. NB096381 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096381 Supplier Novus Biologicals Supplier No. NBP305263

Rabbit Polyclonal Antibody

SIX6 Polyclonal specifically detects SIX6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SIX6
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias homeobox protein SIX6, Homeodomain protein OPTX2, MCOPCT2, Optic homeobox 2, OPTX2, sine oculis homeobox (Drosophila) homolog 6, Sine oculis homeobox homolog 6, sine oculis homeobox homolog 6 (Drosophila), SIX homeobox 6, Six9
Host Species Rabbit
Immunogen Recombinant protein of human SIX6.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4990
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.