Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB031454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB031454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB031454 Supplier Novus Biologicals Supplier No. NBP2835250.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIX6 Polyclonal specifically detects SIX6 in Human samples. It is validated for Western Blot.

Specifications

Antigen SIX6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 2% Sucrose
Gene Alias homeobox protein SIX6, Homeodomain protein OPTX2, MCOPCT2, Optic homeobox 2, OPTX2, sine oculis homeobox (Drosophila) homolog 6, Sine oculis homeobox homolog 6, sine oculis homeobox homolog 6 (Drosophila), SIX homeobox 6, Six9
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the middle region of human SIX6
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4990
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.