Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKI II, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective non-lipid inhibitor of sphingosine kinase (IC50 = 0.5 μM); does not act at ATP-binding site. Displays no inhibition of ERK2, PI 3-kinase, or PKCα at concentrations up to 60 μM. Reduces levels of sphingosine-1-phosphate in MDA-MB-231 breast cancer cells. Induces apoptosis and inhibits proliferation in several other tumor cell lines in vitro (IC50 = 0.9 - 4.6 μM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 302.78 |
| Percent Purity | >98% |
| Chemical Name or Material | SKI II |
Safety and Handling
| Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction