Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Skp2 Rabbit anti-Human, Mouse, Rat, Clone: 2L0K1, Novus Biologicals™
SDP

Catalog No. NB167732 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167732 20 μg
NB167731 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167732 Supplier Novus Biologicals Supplier No. NBP31632020UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Skp2 Monoclonal antibody specifically detects Skp2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Skp2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2L0K1
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CDK2/cyclin A-associated protein p45, cyclin A/CDK2-associated protein p45, Cyclin-A/CDK2-associated protein p45, FBL1, F-box protein Skp2, F-box/LRR-repeat protein 1, FBXL1MGC1366, FLB1, p45skp2, S-phase kinase-associated protein 2, S-phase kinase-associated protein 2 (p45)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Skp2 (Q13309).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6502
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.