Learn More
MSA™ Skullgard™ Protective Caps and Hats
Suitable for use in steel mills
$191.50 - $262.50
Specifications
| Size | Standard (6.5 to 8 in.) |
|---|
Alert:
Do not mix suspension and helmet sizes. Make sure the sizes match (example: standard size suspension with standard size helmet). Use only MSA suspensions on an MSA helmet. Failure to comply with above will reduce the energy-absorbing ability of the protective helmet, which can result in serious injury or death.
| Catalog Number | Mfr. No. | Color | Head Style | Suspension Type | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Color | Head Style | Suspension Type | Price | Quantity & Availability | |||
|
19-043746
|
MSA™
454666 |
Yellow | Full Brim | Staz-On Suspension |
Each for $210.50
|
|
|||
|
18-999-3474
|
MSA™
475396 |
White | Front Brim | Fas-Trac Ratchet |
Each for $212.00
|
|
|||
|
18-999-3494
|
MSA™
475395 |
Natural Tan | Front Brim | Fas-Trac Ratchet |
Each for $204.00
|
|
|||
|
18-999-3441
|
MSA™
460389 |
Natural Tan | Full Brim | Staz-On Suspension |
Each for $262.50
|
|
|||
|
18-999-3506
|
MSA™
454620 |
Red | Front Brim | Staz-On Suspension |
Each for $191.50
|
|
|||
|
18-999-3508
|
MSA™
454623 |
Blue | Front Brim | Staz-On Suspension |
Each for $191.50
|
|
|||
|
18-999-3475
|
MSA™
475401 |
Blue | Front Brim | Fas-Trac Ratchet |
Each for $212.00
|
|
|||
|
19-088-1044
|
MSA™
454668 |
Green | Full Brim | Staz-On Suspension |
Each for $210.50
|
|
|||
|
19-043665
|
MSA™
458899 |
Yellow-Orange | Front Brim | Staz-On Suspension |
Each for $214.50
|
|
|||
|
19-815-467
|
MSA™
454670 |
Blue | Full Brim | Staz-On Suspension |
Each for $210.50
|
|
|||
|
18-999-3463
|
MSA™
454665 |
White | Full Brim | Staz-On Suspension |
Each for $220.00
|
|
|||
|
18-999-3451
|
MSA™
475407 |
Natural Tan | Full Brim | Fas-Trac Ratchet |
Each for $220.00
|
|
|||
|
19-064431
|
MSA™
454621 |
Green | Front Brim | Staz-On Suspension |
Each for $191.50
|
|
|||
|
18-999-3504
|
MSA™
454626 |
Orange | Front Brim | Staz-On Suspension |
Each for $191.50
|
|
|||
|
18-999-3447
|
MSA™
475408 |
White | Full Brim | Fas-Trac Ratchet |
Each for $231.50
|
|
|||
|
19-815-468
|
MSA™
454673 |
Orange | Full Brim | Staz-On Suspension |
Each for $210.50
|
|
|||
|
18-999-3452
|
MSA™
460409 |
Natural Tan | Front Brim | Staz-On Suspension with attached lamp bracket and cord holder |
Each for $249.50
|
|
|||
|
19-043069
|
MSA™
454618 |
White | Front Brim | Staz-On Suspension |
Each for $191.50
|
|
|||
|
18-999-3507
|
MSA™
454622 |
Gray | Front Brim | Staz-On Suspension |
Each for $191.50
|
|
|||
|
18-999-3461
|
MSA™
458702 |
Red-Orange | Front Brim | Staz-On Suspension |
Each for $224.50
|
|
|||
|
18-999-3449
|
MSA™
482002 |
Natural Tan | Front Brim | Fas-Trac Ratchet with attached Welders’ Lugs | N/A | N/A | N/A | ||
Description
Heavy-duty construction makes cap and hat preferred for use in steel mills and other environments that combine elevated temperatures and risk of injury from falling objects.
- Shells are hand-made in the USA of layered, resin-impregnated textiles laminated under pressure
- Wishstands radiant heat loads up to, but not exceeding, 350°F
- Color: Natural tan
Compliance
Third-party certified by SEI to meet the Type I, Class G requirements of ANSI/ISEA Z89.1.
Specifications
| Standard (6.5 to 8 in.) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.