Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SL 327
GREENER_CHOICE

Catalog No. 196910
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-691-0 10 mg
19-691 1 mg
19-695-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-691-0 Supplier Tocris Bioscience™ Supplier No. 1969/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of MEK1 and MEK2; brain penetrant

Selective inhibitor of MEK1 and MEK2 (IC50 values are 0.18 and 0.22 μM respectively); blocks hippocampal LTP in vitro. Brain penetrant in vivo, blocking fear conditioning and learning in rats, and producing neuroprotection in mice, following systemic administration.

Chemical Identifiers

CAS 305350-87-2
Molecular Formula C16H12F3N3S

Specifications

Quantity 10 mg
Formula Weight 335.35
Purity >99%
Molecular Weight (g/mol) 335.348
Chemical Name or Material SL 327
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.