Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLA2 Antibody (Z30P1F12*F4), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345473 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345473 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345473 Supplier Novus Biologicals Supplier No. NBP250293MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLA2 Monoclonal antibody specifically detects SLA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen SLA2
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone Z30P1F12*F4
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias C20orf156, FLJ21992, Modulator of antigen receptor signaling, SLAP-2MARS, SLAP2MGC49845, Src-like adapter protein 2, Src-like adapter protein-2, src-like-adapter 2, Src-like-adaptor 2
Host Species Mouse
Immunogen Peptide Sequence - LRESLSFY
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84174
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.