Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLAIN1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB342234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342234 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342234 Supplier Novus Biologicals Supplier No. NBP281828MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLAIN1 Polyclonal antibody specifically detects SLAIN1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SLAIN1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias SLAIN motif family, member 1
Host Species Rabbit
Immunogen SLAIN1 antibody was raised against an 15 amino acid synthetic peptide near the carboxy terminus of human SLAIN1. The immunogen is located within the last 50 amino acids of SLAIN1. Amino Acid Squence: NGSNLPRSKIAQPVRS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122060
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.