Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLAM/CD150 Antibody (459911), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB347882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347882 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347882 Supplier Novus Biologicals Supplier No. FAB4330MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Mouse samples. It is validated for Flow Cytometry, ELISA Standard (Matched Pair), ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), CyTOF

Specifications

Antigen SLAM/CD150
Applications Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF
Classification Monoclonal
Clone 459911
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SLAM/CD150, Thr25-Pro242, Accession # Q9QUM4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6504
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.