Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLAM/CD150 Antibody (542301) [DyLight 350], Novus Biologicals™
SDP

Catalog No. NB399237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB399237 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB399237 Supplier Novus Biologicals Supplier No. FAB1642D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF

Specifications

Antigen SLAM/CD150
Applications Western Blot, Flow Cytometry, ELISA, CyTOF
Classification Monoclonal
Clone 542301
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, ELISA, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SLAM/CD150, Ala21-Pro237, Accession # Q13291
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6504
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.