Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLAM/CD150 Antibody (MM0553-6L6), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP211907C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211907C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211907C Supplier Novus Biologicals Supplier No. NBP211907C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLAM/CD150
Applications Western Blot
Classification Monoclonal
Clone MM0553-6L6
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen
Gene Symbols SLAMF1
Host Species Mouse
Immunogen Human recombinant SLAM/CD150 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6504
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.