Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLAM/CD150 Antibody (SLAM.4), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP262198A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP262198A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP262198A70 Supplier Novus Biologicals Supplier No. NBP262198AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen SLAM/CD150
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone SLAM.4
Conjugate Alexa Fluor 750
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen
Gene Symbols SLAMF1
Host Species Mouse
Immunogen Human CD150-transfected 300.19 cells
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6504
Test Specificity The antibody MEM-115 recognizes an epitope in the D1 domain of CD4 antigen, a 55 kDa transmebrane glycoprotein expressed on a subset of T lymphocytes and also on monocytes, tissue macrophages and granulocytes. It is negative in Western blotting even with non-reduced samples of cell lysates.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.