Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC10A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18881325UL
Description
SLC10A2 Polyclonal specifically detects SLC10A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC10A2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
Apical sodium-dependent bile acid transporter, ASBT, IBAT, ileal apical sodium-dependent bile acid transporter, Ileal Na(+)/bile acid cotransporter, ileal sodium/bile acid cotransporter, Ileal sodium-dependent bile acid transporter, ISBT, Na(+)-dependent ileal bile acid transporter, NTCP2PBAM, Sodium/taurocholate cotransporting polypeptide, ileal, solute carrier family 10 (sodium/bile acid cotransporter family), member 2, Solute carrier family 10 member 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
6555 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
SLC10A2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NKAEIPESKENGTEPESSFYKANGGFQPDE | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction