Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC10A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SLC10A2 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC10A2 Polyclonal specifically detects SLC10A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC10A2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
6555 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:NKAEIPESKENGTEPESSFYKANGGFQPDE | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Apical sodium-dependent bile acid transporter, ASBT, IBAT, ileal apical sodium-dependent bile acid transporter, Ileal Na(+)/bile acid cotransporter, ileal sodium/bile acid cotransporter, Ileal sodium-dependent bile acid transporter, ISBT, Na(+)-dependent ileal bile acid transporter, NTCP2PBAM, Sodium/taurocholate cotransporting polypeptide, ileal, solute carrier family 10 (sodium/bile acid cotransporter family), member 2, Solute carrier family 10 member 2 | |
SLC10A2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title