Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC12A8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15989320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15989320 20 μL
NBP159893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15989320 Supplier Novus Biologicals Supplier No. NBP15989320UL

Rabbit Polyclonal Antibody

SLC12A8 Polyclonal specifically detects SLC12A8 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC12A8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. A0AV02
Gene Alias CCC9member 8, DKFZp686L18248, FLJ23188, solute carrier family 12 (potassium/chloride transporters), member 8, solute carrier family 12 member 8
Gene Symbols SLC12A8
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC12A8(solute carrier family 12 (potassium/chloride transporters), member 8) The peptide sequence was selected from the N terminal of SLC12A8. Peptide sequence IIRLQLLLLFLLAVSTLDFVVGSFTHLDPEHGFIGYSPELLQ
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84561
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.