Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC13A3/NaDC3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP160011 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP160011 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP160011 Supplier Novus Biologicals Supplier No. NBP160011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC13A3/NaDC3 Polyclonal specifically detects SLC13A3/NaDC3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC13A3/NaDC3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8WWT9
Gene Alias hNaDC3, Na(+)/dicarboxylate cotransporter 3, NaDC-3, NADC3SDCT2naDC-3, sodium-dependent high affinity dicarboxylate transporter 3, Sodium-dependent high-affinity dicarboxylate transporter 2, solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3, solute carrier family 13 member 3
Gene Symbols SLC13A3
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC13A3(solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3) The peptide sequence was selected from the middle region of SLC13A3 (NP_001011554). Peptide sequence GLSFRGWRKNKSEIRTNAEDRARAVIREEYQNLGPIKFAEQAVFILFCMF The peptide sequence for this immunogen was taken from within the described region.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64849
Test Specificity Expected identity based on immunogen sequence: Human: 100%; Canine: 92%; Equine: 92%; Mouse: 92%; Rabbit: 92%; Rat: 92%; Bovine: 85%; Pig: 84%; Chicken: 76%; Zebrafish: 76%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.