Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC14A1 Antibody (888418), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB310939 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310939 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310939 Supplier Novus Biologicals Supplier No. FAB8238PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC14A1 Monoclonal antibody specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen SLC14A1
Applications Flow Cytometry
Classification Monoclonal
Clone 888418
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias FLJ33745, HsT1341, HUT11, JKFLJ41687, RACH1, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1UT-B1, UTEblood group Kidd urea transporter
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC14A1, Accession # Q13336
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6563
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.