Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC15A4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159636 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159636 Supplier Novus Biologicals Supplier No. NBP159636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC15A4 Polyclonal specifically detects SLC15A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC15A4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8N697
Gene Alias Peptide transporter 4, Peptide/histidine transporter 1, PHT1hPHT1, PTR4peptide-histidine transporter 4, solute carrier family 15 member 4, solute carrier family 15, member 4
Gene Symbols SLC15A4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC15A4(solute carrier family 15, member 4) The peptide sequence was selected from the N terminal of SLC15A4 (NP_663623). Peptide sequence MEGSGGGAGERAPLLGARRAAAAAAAAGAFAGRRAACGAVLLTELLERAA.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121260
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.