Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC16A12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen SLC16A12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159530
SDP
View Documents
Novus Biologicals
NBP159530
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

SLC16A12 Polyclonal specifically detects SLC16A12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLC16A12
Polyclonal
Rabbit
Q6ZSM3
387700
IgG
Western Blot
Unconjugated
RUO
CJMG, DKFZp686E188, MCT 12, MCT12monocarboxylate transporter 12, solute carrier family 16 (monocarboxylic acid transporters), member 12, Solute carrier family 16 member 12, solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
SLC16A12
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.