Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC17A9 Antibody, Novus Biologicals™
SDP

Catalog No. NBP230850 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230850 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230850 Supplier Novus Biologicals Supplier No. NBP230850

Rabbit Polyclonal Antibody

SLC17A9 Polyclonal specifically detects SLC17A9 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC17A9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9BYT1
Gene Alias C20orf59, Chromosome 20 Open Reading Frame 59, Solute Carrier Family 17 (Vesicular Nucleotide Transporter), Member 9, Solute Carrier Family 17 Member 9, Solute Carrier Family 17, Member 9, Vesicular Nucleotide Transporter SLC17A9, VNUT
Gene Symbols SLC17A9
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: MQPPPDEARRDMAGDTQWSRPECQAWT
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63910
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.