Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC19A2 Antibody, Novus Biologicals™
SDP

Catalog No. NB176668 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176668 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176668 Supplier Novus Biologicals Supplier No. NBP312981
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC19A2 Polyclonal antibody specifically detects SLC19A2 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen SLC19A2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunofluorescence 1:100-1:1000
Formulation PBS, 20% Glycerol (pH7)
Gene Alias high affinity thiamine transporter, reduced folate carrier protein (RFC) like, solute carrier family 19 (thiamine transporter), member 2, Solute carrier family 19 member 2, TC1, Thiamine carrier 1, thiamine transporter 1, THT1, ThTr1, ThTr-1, THTR1thTr-1, TRMA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC19A2. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10560
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.