Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A4 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB046403 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB046403 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB046403 Supplier Novus Biologicals Supplier No. NBP297677AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC1A4 Polyclonal specifically detects SLC1A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC1A4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Alanine/serine/cysteine/threonine transporter 1, ASCT1, ASCT1 ASCT-1, neutral amino acid transporter A, SATTglutamate/neutral amino acid transporter, solute carrier family 1 (glutamate/neutral amino acid transporter), member 4, Solute carrier family 1 member 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SLC1A4 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6509
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.