Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A5 Antibody (HL1617) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294932 Supplier Novus Biologicals Supplier No. NBP325685
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SLC1A5 Monoclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SLC1A5
Applications Immunocytochemistry
Classification Monoclonal
Clone HL1617
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human SLC1A5. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6510
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.