Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A7 (extracellular) Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5111783
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111783 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111783 Supplier Thermo Scientific Supplier No. PA5111783

Rabbit Polyclonal Antibody

Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)KETTEQS GKPVMS Homology: Rat amino acid residues identical; human - 12/13 amino acid residues identical.

This gene belongs to the sodium:dicarboxylate (SDF) symporter (TC 2.A.23) family, SLC1A7 subfamily. As transportor of L-glutamate; the L-glutamate uptake is sodium- and voltage-dependent and chloride-independent. Its associated chloride conductance may participate in visual processing.It interacts with the PDZ domains of DLG4. Expressed primarily in retina. Detectable in liver, heart, muscle and brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC1A7 (extracellular)
Applications Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.05% sodium azide; pH 7.4
Gene Slc1a7
Gene Accession No. Q8JZR4
Gene Alias A930031E15Rik; AAAT; EAAT5; Excitatory amino acid transporter 5; Retinal glutamate transporter; Slc1a7; Solute carrier family 1 member 7
Gene Symbols Slc1a7
Host Species Rabbit
Immunogen Peptide corresponding to amino acid residues 122-134 of mouse EAAT5 (2nd exracellular loop)
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242607, 366432
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.