Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC22A1 Antibody (609222), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB311688 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311688 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311688 Supplier Novus Biologicals Supplier No. FAB6469APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC22A1 Monoclonal antibody specifically detects SLC22A1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen SLC22A1
Applications Flow Cytometry
Classification Monoclonal
Clone 609222
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias hOCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC22A1, Met1-Thr554, Accession # NP_003048
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6580
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.