Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC22A1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159419 Supplier Novus Biologicals Supplier No. NBP159419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC22A1 Polyclonal specifically detects SLC22A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC22A1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O15245
Gene Alias hOCT1, oct1_cds, OCT1solute carrier family 22 member 1, Organic cation transporter 1, solute carrier family 22 (organic cation transporter), member 1
Gene Symbols SLC22A1
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of human SLC22A1. Peptide Sequence IAIQMICLVNAELYPTFVSGVGPACRGSDATSSRDQGGRFARDHEGRREP. The peptide sequence for this immunogen was taken from within the described region.
Molecular Weight of Antigen 56 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6580
Test Specificity This antibody was created against isoform 2 of the SLC22A1 protein.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.