Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC22A13 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP16252020UL
Description
SLC22A13 Polyclonal specifically detects SLC22A13 in Human samples. It is validated for Western Blot.Specifications
| SLC22A13 | |
| Polyclonal | |
| Western Blot 1:100-1:2000 | |
| Q9Y226 | |
| SLC22A13 | |
| Synthetic peptides corresponding to SLC22A13(solute carrier family 22, member 13) The peptide sequence was selected from the N terminal of SLC22A13. Peptide sequence FFAHVFMVLDEPHHCAVAWVKNHTFNLSAAEQLVLSVPLDTAGHPEPCLM. | |
| Affinity Purified | |
| RUO | |
| 9390 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| PBS and 2% Sucrose with 0.09% Sodium Azide | |
| OAT10, OCTL1, OCTL3, ORCTL3, ORCTL-3, Organic cation transporter-like 3, organic cationic transporter-like 3, organic-cation transporter like 3, solute carrier family 22 (organic anion transporter), member 13, solute carrier family 22 member 13, solute carrier family 22, member 13 | |
| Rabbit | |
| 61 kDa | |
| 20 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction