Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC22A15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP162420
Description
SLC22A15 Polyclonal specifically detects SLC22A15 in Human samples. It is validated for Western Blot.Specifications
SLC22A15 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
SLC22A15 | |
Synthetic peptides corresponding to SLC22A15(solute carrier family 22, member 15) The peptide sequence was selected from the middle region of SLC22A15. Peptide sequence NQKWFGRKRTLSAFLCLGGLACLIVMFLPEKKDTGVFAVVNSHSLSLLGK. | |
Affinity purified | |
RUO | |
55356 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
DKFZp761G0313, Flipt 1, FLIPT1PRO34686, fly-like putative organic ion transporter 1, Fly-like putative transporter 1, solute carrier family 22 (organic cation transporter), member 15, solute carrier family 22 member 15, solute carrier family 22, member 15, trans-like protein | |
Rabbit | |
59 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction