Learn More
Invitrogen™ SLC22A16 Polyclonal Antibody

Description
Immunogen sequence: KLPETLGKRL ATTWEEAAKL ESENESKSSK LLLTTNNSGL EKTEAITPRD SGLGE Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 34%.
Specifications
Specifications
| Antigen | SLC22A16 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SLC22A16 |
| Gene Accession No. | Q86VW1 |
| Gene Alias | Carnitine transporter 2; CT2; dJ261K5.1; epididymis secretory protein Li 18; flipt 2; FLIPT2; fly-like putative organic ion transporter 2; fly-like putative transporter 2; HEL-S-18; OAT6; OCT6; OKB1; organic cation transporter 6; organic cation transporter OKB1; Organic cation/carnitine transporter 6; SLC22A16; solute carrier family 22 (organic cation transporter), member 16; solute carrier family 22 (organic cation/carnitine transporter), member 16; solute carrier family 22 member 16; WUGSC:RG331P03.1 |
| Gene Symbols | SLC22A16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.