Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC22A2/OCT2 Antibody (640438), Biotin, Novus Biologicals™
SDP

Catalog No. NB302997 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302997 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302997 Supplier Novus Biologicals Supplier No. FAB6547B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLC22A2/OCT2 Monoclonal antibody specifically detects SLC22A2/OCT2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry

Specifications

Antigen SLC22A2/OCT2
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 640438
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry
Formulation PBS
Gene Alias hOCT2, MGC32628, OCT2Organic cation transporter 2, solute carrier family 22 (organic cation transporter), member 2, solute carrier family 22 member 2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC-22A2,, Accession # O15244
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6582
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.