Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC22A2/OCT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$382.00 - $646.00
Specifications
Antigen | SLC22A2/OCT2 |
---|---|
Concentration | 0.2mg/mL |
Dilution | Western Blot Reactivity reported in scientific literature (PMID: 26045896)., Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Single Cell Western Single Cell Western reported by an internal validation at a 1:20 dilution |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Description
SLC22A2/OCT2 Polyclonal specifically detects SLC22A2/OCT2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Single Cell Western.Specifications
SLC22A2/OCT2 | |
Western Blot Reactivity reported in scientific literature (PMID: 26045896)., Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Single Cell Western Single Cell Western reported by an internal validation at a 1:20 dilution | |
Polyclonal | |
Rabbit | |
Human, Rat | |
O15244 | |
6582 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ESPRWLISQNKNAEAMRIIKHIAKKNGKSLPASLQRLRLEEETGKKLNPSFLDLVRTPQIR | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
63 kDa |
0.2mg/mL | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
hOCT2, MGC32628, OCT2Organic cation transporter 2, solute carrier family 22 (organic cation transporter), member 2, solute carrier family 22 member 2 | |
SLC22A2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title