Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC22A3 Antibody (SR2204), Novus Biologicals™
SDP

Catalog No. NB260080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260080 100 μg
NB258250 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260080 Supplier Novus Biologicals Supplier No. NBP322312100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SLC22A3 Monoclonal antibody specifically detects SLC22A3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen SLC22A3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone SR2204
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EMTEMTH, Extraneuronal monoamine transporter, OCT3EMT organic cation transporter 3, Organic cation transporter 3, solute carrier family 22 (extraneuronal monoamine transporter), member 3, solute carrier family 22 member 3
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC22A3 (Uniprot #: O75751)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6581
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.