Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC23A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113474
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA5113474 100 μg
1 options

Catalog No. PIPA5113474

Supplier: Invitrogen™ PA5113474

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC23A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Sodium/ascorbate cotransporter. Mediates electrogenic uptake of vitamin C, with a stoichiometry of 2 Na(+) for each ascorbate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC23A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC23A1
Gene Accession No. Q9UHI7, Q9Z2J0
Gene Alias D18Ucla2; hSVCT1; Na(+)/L-ascorbic acid transporter 1; SLC23A1; Slc23a2; Sodium-dependent vitamin C transporter 1; sodium-dependent vitamin C transporter-1; solute carrier family 23 (ascorbic acid transporter), member 1; solute carrier family 23 (nucleobase transporters), member 1; solute carrier family 23 (nucleobase transporters), member 2; solute carrier family 23 member 1; SVCT1; yolk sac permease-like molecule 3; YSPL3
Gene Symbols SLC23A1
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 23 member 1 protein (173-259AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20522, 9963
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.