Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC23A3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15989620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15989620 20 μL
NBP159896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15989620 Supplier Novus Biologicals Supplier No. NBP15989620UL

Rabbit Polyclonal Antibody

SLC23A3 Polyclonal specifically detects SLC23A3 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC23A3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q6PIS1-2
Gene Alias E2-binding protein 3, E2BP3, FLJ31168, Na(+)/L-ascorbic acid transporter 3, Sodium-dependent vitamin C transporter 3, solute carrier family 23 (nucleobase transporters), member 3, solute carrier family 23 member 3, SVCT3HPC E2-binding protein 3, Yspl1
Gene Symbols SLC23A3
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC23A3(solute carrier family 23 (nucleobase transporters), member 3) The peptide sequence was selected from the N terminal of SLC23A3. Peptide sequence SRSPLNPSQLRSVGSQDALAPLPPPAPQNPSTHSWDPLCGSLPWGLSCLL
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151295
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.