Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A12 Antibody (10B12), Novus Biologicals™
SDP

Catalog No. NB298335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298335 100 μL
NB298334 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298335 Supplier Novus Biologicals Supplier No. NBP326321100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SLC25A12 Monoclonal antibody specifically detects SLC25A12 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen SLC25A12
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10B12
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias AGC1, araceli hiperlarga, ARALAR, ARALAR1, calcium binding mitochondrial carrier superfamily member Aralar1, calcium-binding mitochondrial carrier protein Aralar1, Mitochondrial aspartate glutamate carrier 1, solute carrier family 25 (mitochondrial carrier, Aralar), member 12, Solute carrier family 25 member 12
Host Species Rabbit
Immunogen A synthesized peptide derived from Human SLC25A12 [UniProt O75746]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8604
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.