Learn More
Description
Specifications
Specifications
| Antigen | SLC25A13 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.61mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | PBS (pH 7.0)and 20% Glycerol with 0.025% Proclin 300 |
| Gene Alias | ARALAR2CTLN2, calcium-binding mitochondrial carrier protein Aralar2, Citrin, Mitochondrial aspartate glutamate carrier 2, Solute carrier family 25 member 13, solute carrier family 25, member 13 (citrin) |
| Gene Symbols | SLC25A13 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
