Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A18 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317540100UL
This item is not returnable.
View return policy
Description
SLC25A18 Polyclonal antibody specifically detects SLC25A18 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
SLC25A18 | |
Polyclonal | |
Western Blot 0.04 to 0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
GC-2, GC2Glutamate/H(+) symporter 2, mitochondrial glutamate carrier 2, solute carrier family 25 (mitochondrial carrier), member 18, Solute carrier family 25 member 18 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: NDFFRRLLMEDGMQRNLKMEM | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
83733 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction