Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A21 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1595820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1595820 20 μL
NBP159581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1595820 Supplier Novus Biologicals Supplier No. NBP15958120UL

Rabbit Polyclonal Antibody

SLC25A21 Polyclonal specifically detects SLC25A21 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC25A21
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9BQT8
Gene Alias MGC126570, ODC1, ODCmitochondrial 2-oxodicarboxylate carrier, oxodicarboxylate carrier, solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21, Solute carrier family 25 member 21
Gene Symbols SLC25A21
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC25A21(solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21) The peptide sequence was selected from the N terminal of SLC25A21. Peptide sequence FYKGILPPILAETPKRAVKFFTFEQYKKLLGY
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89874
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.