Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A21 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SLC25A21 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP159581
![]() |
Novus Biologicals
NBP159581 |
100 μL |
Each for $487.50
|
|
|||||
NBP1595820
![]() |
Novus Biologicals
NBP15958120UL |
20 μL | N/A | N/A | N/A | ||||
Description
SLC25A21 Polyclonal specifically detects SLC25A21 in Human samples. It is validated for Western Blot.Specifications
SLC25A21 | |
Polyclonal | |
Rabbit | |
Q9BQT8 | |
89874 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
MGC126570, ODC1, ODCmitochondrial 2-oxodicarboxylate carrier, oxodicarboxylate carrier, solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21, Solute carrier family 25 member 21 | |
SLC25A21 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title