Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A25 Antibody, Novus Biologicals™
SDP

Catalog No. NBP159870 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159870 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159870 Supplier Novus Biologicals Supplier No. NBP159870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A25 Polyclonal specifically detects SLC25A25 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC25A25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q6KCM6
Gene Alias APC3, calcium-binding mitochondrial carrier protein SCaMC-2, KIAA1896RP11-395P17.4, MCSC, MCSC3, MGC105138, MGC119514, MGC119515, MGC119516, MGC119517, Mitochondrial ATP-Mg/Pi carrier protein 3, Mitochondrial Ca(2+)-dependent solute carrier protein 3, mitochondrial Ca2+-dependent solute carrier, PCSCL, SCAMC2, SCAMC-2, short calcium-binding mitochondrial carrier 2, small calcium-binding mitochondrial carrier 2, Small calcium-binding mitochondrial carrier protein 2, solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 25, Solute carrier family 25 member 25, solute carrier family 25, member 25
Gene Symbols SLC25A25
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SLC25A25(solute carrier family 25 (mitochondrial carrier, phosphate carrier, member 25). The peptide sequence was selected from the N-terminal of SLC25A25. Peptide sequence AVGLRRLGLHRTEGELQKIVQAGDKDLDGQLDFEEFVHYLQDHEKKLRLV The peptide sequence for this immunogen was taken from within the described region.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114789
Reconstitution Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1mg/ml. Vortex followed by Centrifuge again to pellet the solution.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.