Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A28 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP15956720UL

 View more versions of this product

Catalog No. NBP15956720


Only null left
Add to Cart

Description

Description

SLC25A28 Polyclonal specifically detects SLC25A28 in Human samples. It is validated for Western Blot.
Specifications

Specifications

SLC25A28
Polyclonal
Western Blot 1:100-1:2000
Q96A46
SLC25A28
Synthetic peptides corresponding to SLC25A28(solute carrier family 25, member 28) The peptide sequence was selected from the middle region of SLC25A28. Peptide sequence VWQNEGAGAFYRSYTTQLTMNVPFQAIHFMTYEFLQEHFNPQRRYNPSSH.
20 μL
Primary
Human
IgG
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
DKFZp547C109, hMRS3/4, MFRN2, Mitochondrial iron transporter 2, Mitochondrial RNA-splicing protein 3/4 homolog, mitoferrin-2, MRS3/4MRS4Lmitochondrial RNA splicing protein 3/4, NPD016, putative mitochondrial solute carrier, Solute carrier family 25 member 28, solute carrier family 25, member 28
Rabbit
Affinity Purified
RUO
81894
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.