Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A28 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220390 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220390 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220390 Supplier Novus Biologicals Supplier No. NBP220390

Rabbit Polyclonal Antibody

SLC25A28 Polyclonal specifically detects SLC25A28 in Human samples. It is validated for Western Blot.

Specifications

Antigen SLC25A28
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96A46
Gene Alias DKFZp547C109, hMRS3/4, MFRN2, Mitochondrial iron transporter 2, Mitochondrial RNA-splicing protein 3/4 homolog, mitoferrin-2, MRS3/4MRS4Lmitochondrial RNA splicing protein 3/4, NPD016, putative mitochondrial solute carrier, Solute carrier family 25 member 28, solute carrier family 25, member 28
Gene Symbols SLC25A28
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC25A28. The exact sequence is proprietary.
Molecular Weight of Antigen 39 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81894
Test Specificity Human. Species reactivity based on sequence identity: Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.