Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A31 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $610.00
Specifications
Antigen | SLC25A31 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SLC25A31 Polyclonal specifically detects SLC25A31 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SLC25A31 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
83447 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:FARTRLGVDIGKGPEERQFKGLGDCIMKIAKSDGIAGLYQGFGVSVQGIIVYRA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Human | |
AAC4DKFZp434N1235, adenine nucleotide translocase 4, Adenine nucleotide translocator 4, ADP, ADP/ATP translocase 4, ANT 4, ANT4DKFZP434N1235, ATP carrier protein 4, SFEC, SFEC35kDa, solute carrier family 25 (mitochondrial carrier; adenine nucleotidetranslocator), member 31, Solute carrier family 25 member 31, Sperm flagellar energy carrier protein | |
SLC25A31 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title