Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC25A32 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $499.50
Specifications
Antigen | SLC25A32 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15956620
![]() |
Novus Biologicals
NBP15956620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP159566
![]() |
Novus Biologicals
NBP159566 |
100 μL |
Each for $499.50
|
|
|||||
Description
SLC25A32 Polyclonal specifically detects SLC25A32 in Human samples. It is validated for Western Blot.Specifications
SLC25A32 | |
Polyclonal | |
Rabbit | |
Q9H2D1 | |
81034 | |
Synthetic peptides corresponding to SLC25A32(solute carrier family 25, member 32) The peptide sequence was selected from the middle region of SLC25A32 (NP_110407). Peptide sequence NRLPEAQLSTVEYISVAALSKIFAVAATYPYQVVRARLQDQHMFYSGVID. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MFTCMFTFLJ23872, mitochondrial folate transporter/carrier, Solute carrier family 25 member 32, solute carrier family 25, member 32 | |
SLC25A32 | |
IgG | |
35 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title