Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A33 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132499 Supplier Novus Biologicals Supplier No. NBP132499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A33 Polyclonal specifically detects SLC25A33 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SLC25A33
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9BSK2
Gene Alias BMSC-MCPPNC1 protein, Bone marrow stromal cell mitochondrial carrier protein, HuBMSC-MCP, MGC4399, mitochondrial carrier protein, novel mitochondrial carrier protein, Protein PNC1, solute carrier family 25 member 33, solute carrier family 25, member 33
Gene Symbols SLC25A33
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLC25A33. The exact sequence is proprietary.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84275
Test Specificity Human. Predicted cross-reactivity based on sequence identity: Mouse (88%), Cow (97%), Sheep (98%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.