Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC25A35 Antibody, Novus Biologicals™
SDP

Catalog No. NBP213324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213324 0.1 mL
NB438587 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP213324 Supplier Novus Biologicals Supplier No. NBP213324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC25A35 Polyclonal specifically detects SLC25A35 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SLC25A35
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias FLJ40217, MGC120446, MGC120448, solute carrier family 25 member 35, solute carrier family 25, member 35
Gene Symbols SLC25A35
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: IYMVKTHLQAQAASEIAVGHQYKHQGMFQALTEIGQKHGLVGLWRGALGGLPRVIVGSSTQLCTFSSTKDLLSQ
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399512
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.